Learn More
MilliporeSigma™ anti-RNA Polymerase II, Clone: 8WG16,
Manufacturer:  MilliporeSigma™ 05952
Catalog No. 05-952-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
RNA Polymerase II
8WG16
Mouse monoclonal in buffer containing 0.05% sodium azide and 30% glycerol.
Ascites
Murine
Unpurified
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal
Chromatin Immunoprecipitation, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NP_000928
POLR2A, POLRA, RpIILS, POLR2, RPB1, MGC75453, hsRPB1, RPOL2, RPBh1, RPO2, hRPB220
RNA Polymerase II from wheat germ extract. Clone 8WG16.
100μL
Primary
Human, S. cerevisiae, Yeast