Learn More
RNA polymerase II Mouse anti-Human, Rat, Mouse, Yeast (S. cerevisiae), Clone: CTD4H8, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05623
Catalog No. 05-623-MI
Add to cart
Description
Description
Anti-RNA polymerase II Antibody, clone CTD4H8 is a high quality Mouse Monoclonal Antibody for the detection of RNA polymerase II and has been published in more than 40 citations and validated for use in Chimmunoprecipitation and western blotting.
Specifications
Specifications
RNA polymerase II
CTD4H8
Purified mouse IgG1 in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%.
Purified
Peptide containing 10 repeats of YSPT[pS]PS corresponding to the carboxyl-terminal domain of RNA polymerase II.
Protein G chromatography
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal
Chromatin Immunoprecipitation, Western Blot
Unconjugated
NP_000928
Murine
IgG1
200μg
Primary
Human, Murine, Rat, S. cerevisiae, Yeast