Learn More
MilliporeSigma™ anti-RNA Polymerase II, Clone: CTD4H8, ™ (Upstate™)
Supplier: MilliporeSigma™ 05623/DEL

Description
RNA polymerase II (Pol II) is a multi-subunit enzyme responsible for the transcription of protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. Pol II can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. The DNA binding domain of the polymerase is a groove where TFIIB orients the DNA for unwinding and transcription. GenBank Number: NM_000937. MW: 210-220kDa.
Chromatin Immunoprecipitation, Western Blotting
Specifications
RNA Polymerase II | |
Unconjugated | |
Peptide containing 10 repeats of YSPT[pS]PS corresponding to the carboxyl-terminal domain of RNA polymerase II | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
200 μg | |
Epigenetics | |
Human, Mouse, Rat, Yeast | |
IgG1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.