Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA Polymerase II/POLR2A, p Thr4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP149546
Description
RNA Polymerase II/POLR2A Polyclonal specifically detects RNA Polymerase II/POLR2A in Human, Chicken, Drosophila, Yeast samples. It is validated for Western Blot, Dot Blot.Specifications
RNA Polymerase II/POLR2A (p Thr4) | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for the phosphorylated Threonine found in position 4 of the RNA polymerase II CTD heptad. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Dot Blot | |
1.1 mg/mL | |
Western Blot 1:1000, Dot Blot 1:1000 | |
P24928 | |
POLR2A | |
A synthetic peptide made to a phosphorylated Threonine (position 4) of the CTD heptad in the human RNA polymerase II protein [UniProt P24928] | |
0.1 mL | |
Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators | |
5430 | |
Human, Chicken, Drosophila, Yeast | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction