Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA polymerase II subunit B1 Rat anti-Mouse, Human, Clone: 4F8, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma 041569
Description
Anti-RNA polymerase II subunit B1 Antibody, clone 4F8 is a Rat monoclonal antibody for detection of RNA polymerase II subunit B1 also known as DNA-directed RNA polymerase II A and has been validated in western blot, ELISA.Specifications
RNA polymerase II subunit B1 | |
Monoclonal | |
Unconjugated | |
NP_000928 | |
Rat | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
ELISA, Western Blot | |
4F8 | |
Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
POLR2A, POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1 | |
Ovalbumin-conjugated linear peptide corresponding to human RNA polymerase II subunit B1. | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only