Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA polymerase II subunit B1 (phospho CTD Ser-2), Rat anti-Mouse, Human, Clone: 3E1, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma 041571I
Description
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2), clone 3E1 (rat monoclonal) is a highly specific rat monoclonal antibody and that targets RNA Polymerase and has been tested in western blotting.Specifications
RNA polymerase II subunit B1 (phospho CTD Ser-2) | |
Monoclonal | |
Unconjugated | |
NP_000928 | |
Rat | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Western Blot | |
3E10 | |
Unpurified rat monoclonal IgG1κ with 0.05% sodium azide. | |
POLR2A, POLR2 | |
Ovalbumin-conjugated linear peptide corresponding to human RNA Polymerase II subunit B1 phosphorylated at Ser2 of the C-terminal domain (CTD). | |
100 μL | |
Human, Mouse | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only