Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA polymerase II subunit B1 (phospho-CTD Ser-7) Rat anti-Mouse, Clone: 4E12, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma 041570
Description
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12 is a Rat monoclonal antibody for detection of RNA polymerase II subunit B1 (phospho-CTD Ser-7) has been validated in western blot, ELISA.Specifications
RNA polymerase II subunit B1 (phospho-CTD Ser-7) | |
Monoclonal | |
Unconjugated | |
NP_000928 | |
Rat | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
ELISA, Western Blot | |
4E12 | |
Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
POLR2A, POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1 | |
Ovalbumin-conjugated linear peptide corrresponding to human RNA polymerase subunit B1 CTD phosphorylated at Ser7. | |
100 μg | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only