Learn More
anti-RNASEH2A, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP176981
Catalog No. NBP176981
Add to cart
Description
Description
RNASEH2A Polyclonal antibody specifically detects RNASEH2A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.
Specifications
Specifications
RNASEH2A
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation, Immunofluorescence 4 ug/ml, Immunocytochemistry 2 ug/ml
O75792
AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL
Rabbit
IgG
Peptide affinity purified
RUO
Primary
10535
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
RNASEH2A
Antibody was raised against a 17 amino acid synthetic peptide near the center of human RNAse H2A. The immunogen is located within amino acids 190 - 240 of RNAse H2A.
33 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat