Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RNASEH2A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232143

Catalog No. NBP232143


Add to cart

Description

Description

RNASEH2A Polyclonal antibody specifically detects RNASEH2A in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

RNASEH2A
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
NP_006388.2
AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
RNASEH2A
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ribonuclease H2, Subunit A using the numbering given in entry NP_006388.2 (GeneID 10535).
Immunogen affinity purified
RUO
Primary
10535
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only