Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNaseK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525183

Catalog No. PIPA525183

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and mouse based on sequence homology.

RNASEK is endoribonuclease which preferentially cleaves ApU and ApG phosphodiester bonds. RNASEK hydrolyzes UpU bonds at a lower rate.
Specifications

Specifications

RNASEK
Polyclonal
PBS with 0.09% sodium azide
Q6P5S7
RNase kappa, ribonuclease kappa
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
440400
Immunohistochemistry, Western Blot
Unconjugated
RNASEK
Liquid
RNASEK
KLH conjugated synthetic peptide between 26-55 amino acids from the central region of human RNASEK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit