Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho family GTPase 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RND3 protein.

Manufacturer:  Abnova Corporation H00000390B02

Catalog No. 89-004-929


Add to cart

Description

Description

Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras (MIM 190020), Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex (see MIM 601925); GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins (see MIM 601855); and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form (see MIM 602680) (Nobes et al., 1998 [PubMed 9531558]).[supplied by OMIM

Sequence: MKERRASQKLSSKSIMDPNQNVKCKIVVVGDSQCGKTALLHVFAKDCFPENYVPTVFENYTASFEIDTQRIELSLWDTSGSPYYDNVRPLSYPDSDAVLICFDISRPETLDSVLKKWKGEIQEFCPNTKMLLVGCKSDLRTDVSTLVELSNHRQTPVSYDQGANMAKQIGAATYIECSALQSENSVRDIFHVATLACVNKTNKNVKRNKSQRATKRISHMPSRPELSAVATDLRKDKAKSCTVM
Specifications

Specifications

Rho family GTPase 3
Unconjugated
No additive
NM_005168
ARHE|Rho8|RhoE|memB
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human RND3 protein.
RND3
Serum
RND3
RND3 (NP_005159, 1 a.a. ~ 244 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
390
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit