Learn More
RNF113B Mouse anti-Human, Clone: 2B11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA523047
Catalog No. PIMA523047
Add to cart
Description
Description
Peptide Sequence: MAAPPSPGRT ADQADQVCTF LFKKPGRKGA AGLRKRPACD PEHGESSSSG DEGDTVAQPP RVAPRPRGLH SWQKAAHGDR RGEEAAPESL DVVYRSTRSA
Specifications
Specifications
RNF113B
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8IZP6
RNF161, ZNF183L1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
140432
ELISA, Immunoprecipitation, Western Blot
2B11
Unconjugated
RNF113B
Liquid
RNF113B
RNF113B (NP_849192, 1 a.a. ∽ 100 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human