Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124404

Catalog No. PIPA124404


Add to cart

Description

Description

PA1-24404 detects RNF12 in human samples. PA1-24404 has been successfully used in Western blot and immunohistochemistry procedures. The PA1-24404 immunogen is a synthetic peptide corresponding to a region of human RNF12. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

RNF12 is a RING-H2 zinc finger protein. It has been shown to be a ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Alternatively spliced transcript variants encoding the same protein have been reported. Transcript Variant: This variant (1) lacks an exon in the 5 ft. UTR compared to variant 2, and may represent the more abundant form. Both variants encode the same protein.
Specifications

Specifications

RNF12
Polyclonal
Unconjugated
RLIM
Liquid
RLIM
Synthetic peptide corresponding to a region of human RNF12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9NVW2
CTD-2530H13.3, DKFZp686N06224, FLJ25923, FLJ41093, FLJ42887, FLJ45986, MGC15161, NY-REN-43, RNF12
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51132
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit