Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RNF139, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP123027

Catalog No. NBP123027


Add to cart

Description

Description

RNF139 Polyclonal antibody specifically detects RNF139 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

RNF139
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Immunohistochemistry 3 - 4 ug/ml, Immunohistochemistry-Paraffin 3 - 4 ug/ml, Peptide ELISA Detection limit 1:16000
NP_009149.2
EC 6.3.2.-, HRCA1, patched related protein translocated in renal cancer, RCA1multiple membrane spanning receptor TRC8, ring finger protein 139MGC31961, Translocation in renal carcinoma on chromosome 8 protein, TRC8E3 ubiquitin-protein ligase RNF139
Goat
IgG
0.1 mg
Zinc Finger
Primary
11236
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
RNF139
Peptide with sequence C-HTGAAAEEFNDDT corresponding to C-Terminus according to NP_009149.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Human. Expected from sequence similarity: Dog.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only