Learn More
MilliporeSigma™ anti-RNF168 Ab 2, Polyclonal,
Manufacturer:  MilliporeSigma™ 061130
Catalog No. 06-113-0MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
RNF168
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Immunoprecipitation, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
RNF168
Histidine-tagged recombinant protein corresponding to the C-terminus of human RNF168.
100μg
Primary
NP_689830