Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF183 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523862

Catalog No. PIPA523862


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

RNF183
Polyclonal
PBS with 0.09% sodium azide
Q96D59, Q8QZS5
RING finger protein 183
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
138065, 76072
Western Blot
Unconjugated
RNF183
Liquid
RNF183
KLH conjugated synthetic peptide between 86-114 amino acids from the central region of human RNF183
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit