Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF208 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525646

Catalog No. PIPA525646

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RNF208
Polyclonal
PBS with 0.09% sodium azide
Q9H0X6, Q8K0W3
RING finger protein 208
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
68846, 727800
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
RNF208
Liquid
RNF208
KLH conjugated synthetic peptide between 86-115 amino acids from the central region of human RNF208
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit