Learn More
tripartite motif-containing 69, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RNF36 protein.
Manufacturer: Abnova Corporation H00140691B01
Description
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Sequence: MEEELAIQQGQLETTLKELQTLRNMQKEAIAAHKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSLEQGMKVLATRELISRKLNLGQYKGPIQYMVWREMQDTLCPGLSPLTLDPKTAHPNLVLSKSQTSVWHGDIKKIMPDDPERFDSSVAVLGSRGFTSGKWYWEVEVAKKTKWTVGVVRESIIRKGSCPLTPEQGFWLLRLRNQTDLKALDLPSFSLTLTNNLDKVGIYLDYEGGQLSFYNAKTMTHIYTFSNTFMEKLYPYFCPCLNDGGENKEPLHILHPQSpecifications
tripartite motif-containing 69 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human RNF36 protein. | |
TRIM69 | |
HSD34/RNF36/Trif | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_080745.3 | |
TRIM69 | |
RNF36 (NP_542783.2, 1 a.a. ∼ 341 a.a) full-length human protein. | |
RUO | |
Yes | |
140691 | |
Serum |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.