Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF36 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512214
Description
TRIM69 is a member of the RING-B-box-coiled-coil (RBCC) family with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of TRIM69 is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis.Specifications
RNF36 | |
Polyclonal | |
TRIM69 | |
Tripartite motif-containing protein 69; RING finger protein 36; RFP-like domain-containing protein trimless; RNF36; HSD34; HSD-34 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
140691 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q86WT6 | |
TRIM69 | |
KLH conjugated synthetic peptide between 200-231 amino acids from the central region of human RNF36 (TRIM69) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction