Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526820

Catalog No. PIPA526820

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RNF8 contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins.
Specifications

Specifications

RNF8
Polyclonal
PBS with 0.09% sodium azide
O76064
C3HC4-type zinc finger protein, E3 ubiquitin-protein ligase RNF8, UBC13/UEV-interacting ring finger protein, ring finger protein (C3HC4 type) 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9025
Flow Cytometry, Western Blot
Unconjugated
RNF8
Liquid
RNF8
recombit protein of human RNF8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit