Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNaseH2C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526311

Catalog No. PIPA526311


Add to cart

Description

Description

RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.
Specifications

Specifications

RNaseH2C
Polyclonal
Unconjugated
RNASEH2C
Liquid
RNASEH2C
KLH conjugated synthetic peptide between 80-108 amino acids from the central region of human RNH2C
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.41 mg/mL
PBS with 0.09% sodium azide
Q8TDP1
AGS3, AYP1, RNase H1 small subunit, RNase H2 subunit C, aicardi-Goutieres syndrome 3 protein, ribonuclease H2 subunit C, ribonuclease HI subunit C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84153
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit