Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASET2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524253

Catalog No. PIPA524253


Add to cart

Description

Description

This ribonuclease gene is a novel member of the Rh/T2/S-glycoprotein class of extracellular ribonucleases. It is a single copy gene that maps to 6q27, a region associated with human maligcies and chromosomal rearrangement.
Specifications

Specifications

RNASET2
Polyclonal
PBS with 0.09% sodium azide
O00584
RNASE6PL, bA514O12.3, ribonuclease 6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8635
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RNASET2
Liquid
RNASET2
KLH conjugated synthetic peptide between 10-36 amino acids from the N-terminal region of human RNT2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit