Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
roundabout, axon guidance receptor, homolog 3 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ROBO3 protein.
Supplier: Abnova Corporation H00064221B02
Description
Sequence: MTPPLQGPRARFRKKPKALPYRRENSPGDLPPPPLPPPEEEASWALELRAAGSMSSLERERSGERKAVQAVPLAAQRVLHPDEEAWLPYSRPSFLSRGQGTSTCSTAGSNSSRGSSSSRGSRGPGRSRSQSRSQSQRPGQKRREEPRSpecifications
roundabout, axon guidance receptor, homolog 3 (Drosophila) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ROBO3 protein. | |
ROBO3 | |
FLJ21044/HGPPS/HGPS/RBIG1/RIG1 | |
Mouse | |
50 μL | |
Neurobiology | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC008623 | |
ROBO3 | |
ROBO3 (AAH08623, 1 a.a. ∼ 147 a.a) full-length human protein. | |
RUO | |
Yes | |
64221 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title