Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146010

Catalog No. PA146010


Add to cart

Description

Description

In Western Blot, bands at 85 kDa and 107 kDa are observed, representing different isoforms of ROBO4 protein. Suggested positive control: HUVEC lysate, antigen standard for ROBO4 (transient overexpression lysate).

Roundabouts (ROBO) are cell-surface receptors that mediate repulsive signaling mechanisms at the central nervous system midline. However, ROBOs may also mediate attraction mechanisms in the context of vascular development. ROBO4 is a novel roundabout protein which is restricted in expression to endothelial cells in vitro and sites of angiogenesis in vivo. Because it is not expressed on normal endothelial cells, in vivo, it is a promising tumor endothelial marker.
Specifications

Specifications

ROBO4
Polyclonal
Unconjugated
ROBO4
Liquid
ROBO4
Synthetic peptide made to a C-terminal region of the human protein (within residues 900-1007).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
tris citrate with 0.1% sodium azide; pH 7-8
Q8WZ75
ECSM4, FLJ20798, MGC133352, MGC133353, MRB
Rabbit
IgG
100 μL
4° C, do not freeze
Primary
54538
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit