Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK-I Mouse, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611136

 View more versions of this product

Catalog No. BDB611136


Add to cart

Description

Description

ROCK-I is a Rho-associated serine/threonine kinase isozyme that mediates RhoA-induced assembly of focal adhesions and actin stress fibers. It contains an N-terminal kinase domain, a central 600 amino acid long coiled-coil region, a C-terminal pleckstrin homology region (PH) and a Cys-rich zinc finger motif. The ROCK-I kinase domain is approximately 90% identical to that of ROCK-II. ROCK-I binds GTP-bound Rho through a Rho-binding domain (RBD). As a result, the kinase activity of ROCK-I is moderately stimulated. The ROCK isozymes regulate cell contractility through phosphorylation of the myosin light chain. This effect results from either the inhibition of the myosin phosphatase or by direct phosphorylation of the myosin light chain, thus bypassing the myosin light chain kinase. In addition, ROCK-I activates the ubiquitously expressed Na-H exchanger (NHE1) via a number of mechanisms including RhoA. NHE1 may mediate ROCK-I-induced changes in the actin cytoskeleton. Therefore, ROCK-I plays an important role in the regulation of focal adhesion and stress fiber formation.

Host Species: Mouse
Clone: 46
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse ROCK-I aa. 906-1012
Formula Weight [Chemical]: 160kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

ROCK-I
46
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
ROKβ
Affinity Purified
Mouse ROCK-I aa. 906-1012
50μg
Cell Biology
Primary
Human, Murine, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.