Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK-II Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

$232.00 - $458.00

Specifications

Antigen ROCK-II
Isotype IgG1
Immunogen Rat ROKα aa. 567-718
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610624
View Documents Promotion Details
BD Biosciences
610624
150μg
Each for $458.00
Add to cart
 
BDB610623
View Documents Promotion Details
BD Biosciences
610623
50μg
Each for $232.00
Add to cart
 
Description

Description

Activity of the GTP-binding proteins is regulated by GAPs, that accelerate binding, and GTPases, that enhance the rate of GTP hydrolysis (Ras, Rho, cdc42Hs, and Rac). The isoforms of Rho, a small GTP-binding protein, regulate cellular processes such as the formation of the stress fibers, lamelipodia, and filopodia. ROKα (RhoA-binding kinase) is a Ser/Thr protein kinase that interects with the GTP-binding form of RhoA. Like RhoA, ROKα localizes at the cellular margins and colocalizes with acti filaments. The ROKα gene encodes a protein of 1302 amino acids with homology to the human myotonic kinase. Although ROKα binds to GTP-RhoA, it is not yet clear whether this interaction induces the kinase activity of ROKα.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

ROCK-II
Rat ROKα aa. 567-718
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
ROKα
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
21
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit