Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RORA Rabbit anti-Bovine, Chicken, Equine, Human, Marsupial, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523267

Catalog No. PIPA523267

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).
Specifications

Specifications

RORA
Polyclonal
Unconjugated
RORA
Liquid
LOC100147460, RORA
Synthetic peptide (238 QTFLQEEIENYQNKQR 253) of ROR alpha protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Chicken, Equine, Human, Marsupial, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P35398, P51448
NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA, ROR-alpha, nuclear receptor ROR-alpha, nuclear receptor RZR-alpha, nuclear receptor subfamily 1 group F member 1, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, thyroid hormone nuclear receptor alpha variant 4, transcription factor RZR-alpha
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100147460, 19883, 415377, 535597, 6095
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit