Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ ROR gamma T Mouse anti-Human, Clone: 6F3.1
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABF81
Description
This Anti-ROR gamma T Antibody, clone 6F3.1 is validated for use in western blot, immunohistochemistry for the detection of ROR gamma T.
Specifications
ROR gamma T | |
Monoclonal | |
Unconjugated | |
NP_001001523 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunohistochemistry, Western Blot | |
6F3.1 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
RORC, NR1F3, RORG, RZRG | |
GST-tagged recombinant protein corresponding to human ROR gamma T. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction