Learn More
ROR2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06018
Catalog No. 06-018-MI
Add to cart
Description
Description
Anti-ROR2 Antibody, C-terminus detects level of ROR2 and has been published and validated for use in immunohistochemistry (paraffin).
Specifications
Specifications
ROR2
Unconjugated
Purified
Rabbit
Protein G purfied
2-8°C for 12 months
Polyclonal
Human
Immunohistochemistry (Paraffin)
Purified rabbit polyclonal in buffer containing PBS and 0.09% sodium azide.
BDB; BDB1 ; EC 2.7.10.1 ; MGC163394; NTRKR2; OTTHUMP00000021634; OTTHUMP00000063680
KLH-conjugated synthetic linear peptide corresponding to the C-terminal region of ROR2
100μg
Primary
NP_004551.2