Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527991

Catalog No. PIPA527991

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a receptor protein tyrosine kinase and type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors. The protein may be involved in the early formation of the chondrocytes and may be required for cartilage and growth plate development. Mutations in this gene can cause brachydactyly type B, a skeletal disorder characterized by hypoplasia/aplasia of distal phalanges and nails. In addition, mutations in this gene can cause the autosomal recessive form of Robinow syndrome, which is characterized by skeletal dysplasia with generalized limb bone shortening, segmental defects of the spine, brachydactyly, and a dysmorphic facial appearance.

PA5-27991 targets ROR2 in WB applications and shows reactivity with Human samples.The PA5-27991 immunogen is recombinant fragment corresponding to a region within amino acids 353 and 664 of Human ROR2.
Specifications

Specifications

ROR2
Polyclonal
Unconjugated
ROR2
Purified
ROR2
Recombinant fragment corresponding to a region within amino acids 353 and 664 of Human ROR2
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
4920
Western Blot
1mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q01974
BDB, BDB1, NTRKR2, neurotrophic tyrosine kinase receptor-related 2, tyrosine-protein kinase transmembrane receptor ROR2
Rabbit
IgG
100μL
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit