Learn More
ROBO4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525023
Catalog No. PIPA525023
Add to cart
Description
Description
Roundabout 4 is receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. This protein may mediate the inhibition of primary endothelial cell migration by Slit proteins.
Specifications
Specifications
ROBO4
Polyclonal
Unconjugated
ROBO4
Liquid
ROBO4
KLH conjugated synthetic peptide between 236-264 amino acids from the N-terminal region of human Roundabout 4
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8WZ75
ECSM4, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54538