Learn More
Aviva Systems Biology Corp ANTI-RP13-102H20.1 50UG/50ULA DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP52767_P050
This is a rabbit polyclonal antibody against RP13-102H20.1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RP13-102H20.1. Alternate names: FLJ30058, RP13-102H20.1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Pig, Sheep
Catalog No. 50-014-53835