Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPA1 p70 Mouse anti-Human, Clone: RPA9, 1-30, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041521
Description
Use Anti-RPA1 p70 Antibody, clone RPA9, 1-30 (mouse monoclonal antibody) validated in western blot, immunocytochemistry to detect RPA1 p70 also known as RF-A protein 1, RP-A p70, Replication factor A protein 1, REPA1, RPA1, HSSB.Specifications
RPA1 p70 | |
Monoclonal | |
Unconjugated | |
NM_002945 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
RPA9, 1-30, 1-30 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
RPA1, REPA1, RF-A, RP-A, RPA70 | |
His-tagged recombinant protein corresponding to human RPA1. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only