Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLR3H Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524861
Description
RPC8 is DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It is plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. It is acting as nuclear and cytosolic DNA sensor involved in innate immune response. It can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF-Kappa-B through the RIG-I pathway.Specifications
POLR3H | |
Polyclonal | |
POLR3H | |
RPC22.9, RPC8, DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide, DNA-directed RNA polymerase III subunit H, DNA-directed RNA polymerase III subunit RPC8, RNA nucleotidyltransferase (DNA-directed), RNA polymerase III subunit 22.9 kDa subunit, RNA polymerase III su | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171568, 78929 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q9Y535, Q9D2C6 | |
POLR3H | |
KLH conjugated synthetic peptide between 5-33 amino acids from the N-terminal region of human RPC8 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction