Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL13A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521430

Catalog No. PIPA521430


Add to cart

Description

Description

PA5-21430 targets RPL13A in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21430 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 203 of RPL13A (Uniprot IDNo.P40429). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

RPL13A
Polyclonal
Unconjugated
RPL13A
Liquid
RPL13A
Recombit fragment corresponding to a region within amino acids 1 and 203 of RPL13A (Uniprot IDNo.P40429)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.72 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P40429
L13A, TSTA1, 23 kDa highly basic protein, 60S ribosomal protein L13a, tissue specific transplantation antigen 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23521
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit