Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL17 Mouse anti-Human, Mouse, Clone: 702142, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524323

Catalog No. PIMA524323


Add to cart

Description

Description

This antibody detects human Ribosomal Protein L17 in direct ELISAs, and human and mouse Ribosomal Protein L17 in Western blots. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L22P family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as rpL23 because the encoded protein shares amino acid identity with ribosomal protein L23 from Halobacterium marismortui, however, its official symbol is RPL17. Two alternative splice variants have been observed, each encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications

Specifications

RPL17
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P18621, Q9CPR4
L17, PD-1, RPL23, 60S ribosomal protein L17, 60S ribosomal protein L23, gene encoding putative NFkB activating protein
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
319195, 6139
Immunohistochemistry (Paraffin), Western Blot
702142
Unconjugated
RPL17
Lyophilized
RPL17
E. coli-derived recombit human Ribosomal Protein L17 Cys70-Glu184
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit