Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL31 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526540

Catalog No. PIPA526540


Add to cart

Description

Description

This antibody is predicted to react with bovine, mouse, porcine, rat and Xenopus based on sequence homology.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL31 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L31E family of ribosomal proteins. It is located in the cytoplasm.
Specifications

Specifications

RPL31
Polyclonal
PBS with 0.09% sodium azide
P62899
L31, 60S ribosomal protein L31
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6160
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
RPL31
Liquid
RPL31
KLH conjugated synthetic peptide between 24-51 amino acids from the central region of human RPL31
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit