Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPRD1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526915

Catalog No. PIPA526915

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Specifications

Specifications

RPRD1B
Polyclonal
PBS with 0.09% sodium azide
Q9NQG5
C20orf77, CREPT, NET60, dJ1057B20.2, cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, regulation of nuclear pre-mRNA domain-containing protein 1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
58490
Flow Cytometry, Western Blot
Unconjugated
RPRD1B
Liquid
RPRD1B
KLH conjugated synthetic peptide between 254-282 amino acids from the C-terminal region of human RPR1B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit