Learn More
RPS6KB2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526010
Catalog No. PIPA526010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
RPS6KB2 is a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates the S6 ribosomal protein and eucaryotic translation initiation factor 4B. Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
Specifications
RPS6KB2
Polyclonal
Unconjugated
RPS6KB2
Liquid
RPS6KB2
human RPS6KB2 recombit protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry, Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q9UBS0, Q9Z1M4
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb, 70 kDa ribosomal protein S6 kinase 2, P70S6K2, S6 kinase-related kinase, S6K-beta, S6K-beta-2, p70 S6 kinase beta, p70 S6K-beta, p70 S6KB, p70 ribosomal S6 kinase beta, p70-S6K 2, ribosomal protein S6 kinase beta-2, serine/threonine-protein kinase 14 beta, serine/thr
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
58988, 6199