Learn More
RPS9 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA513569
Catalog No. PIPA513569
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS9 is a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S4P family of ribosomal proteins. It is located in the cytoplasm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPS9
Polyclonal
RPS9
40S ribosomal protein S9; RPS9
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
6203
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
P46781
RPS9
KLH conjugated synthetic peptide between 135-164 amino acids from the C-terminal region of human RPS9
400 μL
Primary
Human
Antibody
IgG
Recommendations
Recommendations