Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPTPα Mouse anti-Human, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610348

Catalog No. BDB610348


Add to cart

Description

Description

Receptor-like protein tyrosine phosphatases (RPTPs) represent an important pathway for signal transduction via modification of protein tyrosine phosphorylation status. RPTPα is widely expressed and is particularly abundant in brain. Structurally, it is typical of RPTPs in that it has an intracellular region containing two tandem catalytic domains linked to a transmembrane and an extracellular region. However, unlike many RPTPs, the extracellular region of RPTPα is small (123 residues) and lacks any obvious structural motifs, though the protein is known to be heavily glycosylated. Both catalytic domains of RPTPα have intrinsic in vitro activity, but their exact roles in vivo remain uncertain. It has been postulated that due to their transmembrane nature, RPTPs are initiators for a cascade of intracellular signaling events, much like receptor tyrosine kinases.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

RPTPα
21
Unconjugated
Affinity Purified
Human RPTPα aa. 244-503
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgM
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit