Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-RRAGC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232084

Catalog No. NBP232084



Description

Description

RRAGC Polyclonal antibody specifically detects RRAGC in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

RRAGC
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
NP_071440.1
FLJ13311, GTPase-interacting protein 2, GTR2, Rag C, Rag C protein, RAGC, Ras-related GTP binding C, ras-related GTP-binding protein C, TIB929
Rabbit
IgG
100 ul
Apoptosis, mTOR Pathway
Primary
64121
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
RRAGC
The immunogen this antibody was made to, maps to a region between residue 349 to 399 of human Ras-Related GTP Binding C using the numbering given in entry NP_071440.1 (GeneID 64121).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only