Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RRAGD, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232106

Catalog No. NBP232106


Add to cart

Description

Description

RRAGD Polyclonal antibody specifically detects RRAGD in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

RRAGD
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
NP_067067.1
bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D
Rabbit
IgG
100 ul
Cancer, mTOR Pathway
Primary
58528
Human, Mouse
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
RRAGD
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ras-Related GTP Binding D using the numbering given in entry NP_067067.1 (GeneID 58528).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Gorilla,Chimpanzee, Northern white-cheeked gibbon and Small-eared galago.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only