Learn More
RRH Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00148W
Catalog No. PIOSR00148W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
May play a role in rpe physiology either by detecting light directly or by monitoring the concentration of retinoids or other photoreceptor-derived compounds. Found only in the eye, where it is localized to the retinal pigment epithelium (RPE). In the RPE, it is localized to the microvilli that surround the photoreceptor outer segments.
Specifications
Specifications
Rrh
Polyclonal
whole serum with no preservative
O35214
Visual pigment-like receptor peropsin, mouse retinal pigment epithelium derived rhodopsin homolog OPSX
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Rrh
Lyophilized
RRH
A synthetic peptide from mouse peropsin
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20132