Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

RRM2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127760

Catalog No. PIPA127760

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provides the precursors necessary for DNA synthesis. Catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. Ribonucleotide reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. It is composed o

PA1-27760 detects RRM2 from human samples.PA1-27760 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Ribonucleoside-diphosphate reductase M2 subunit-.
Specifications

Specifications

RRM2
Polyclonal
PBS with no preservative
P31350
R2, RR2, RR2M
Chicken
IgY
50μg
Store at -20°C
Primary
6241
Western Blot
Unconjugated
RRM2
Purified
RRM2
Synthetic peptide corresponding to human Ribonucleoside-diphosphate reductase M2 subunit-.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit