Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521699

Catalog No. PIPA521699


Add to cart

Description

Description

PA5-21699 targets RSK1 p90 in IF and WB applications and shows reactivity with Human samples. The PA5-21699 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 303 and 527 (Q15418) of RSK1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

RSK1
Polyclonal
Unconjugated
RPS6KA1
Liquid
RPS6KA1
Recombit protein fragment contain a sequence corresponding to a region within amino acids 303 and 527 (Q15418) of RSK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.47 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q15418
RP11-492M19.2, HU-1, MAPKAPK1A, RSK, RSK1, 90 kDa ribosomal protein S6 kinase 1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, RSK-1, S6K-alpha 1, S6K-alpha-1, dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1), p90-RSK 1, p90RSK1, p90S6K, ribosomal S6 kinase 1, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6195
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit