Learn More
TSGA2 Mouse anti-Human, Clone: 3G6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522825
Catalog No. PIMA522825
Add to cart
Description
Description
Peptide Sequence: EELVTVVPKW KATQITELAL WTPTLPKKPT STDGPGQDAP GAESAGEPGE EAQALLEGFE GEMDMRPGDE DADVLREESR EYDQEEFRYD MDEGNINSEE EETRQSDLQD
This gene encodes a male meiotic metaphase chromosome-associated acidic protein. This gene is expressed in tissues with motile cilia or flagella, including the trachea, lungs, airway brushings, and testes. Mutations in this gene result in primary ciliary dyskinesia-24. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2014]
Specifications
Specifications
TSGA2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8WYR4
ABCB2, PSF1, RING4, Y3
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
89765
ELISA, Western Blot
3G6
Unconjugated
RSPH1
Liquid
RSPH1
RSPH1 (NP_543136, 200 a.a. ∽ 309 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human