Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSPO4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525615

Catalog No. PIPA525615


Add to cart

Description

Description

RSPO4 is a member of the R-spondin family of proteins that share a common domain organization consisting of a signal peptide, cysteine-rich/furin-like domain, thrombospondin domain and a C-terminal basic region. The encoded protein may be involved in activation of Wnt/beta-catenin signaling pathways. Mutations in this gene are associated with anonychia congenital. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

RSPO4
Polyclonal
PBS with 0.09% sodium azide
Q2I0M5
C20orf182, CRISTIN4, R-spondin family, member 4, R-spondin-4, hRspo4, roof plate-specific spondin-4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
343637
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RSPO4
Liquid
RSPO4
KLH conjugated synthetic peptide between 199-228 amino acids from the C-terminal region of human RSPO4
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit