Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS34 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524471

Catalog No. PIPA524471


Add to cart

Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated75% protein to rRNA composition compared to prokaryotic ribosomes,where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. Alternate splice variants of this gene have been described but their full-length nature has not been determined.
Specifications

Specifications

MRPS34
Polyclonal
Unconjugated
MRPS34
Liquid
MRPS34
KLH conjugated synthetic peptide between 4-32 amino acids from the N-terminal region of human RT34
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P82930, Q9JIK9
MRP-S12, MRP-S34, MRPS12, 28S ribosomal protein S34, mitochondrial, S34mt
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65993, 79044
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit