Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524995

Catalog No. PIPA524995

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RTP2 specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
Specifications

Specifications

RTP2
Polyclonal
PBS with 0.09% sodium azide
Q5QGT7, Q80ZI2
receptor transporter protein 2, receptor transporting protein 2, receptor-transporting protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
224055, 344892
Western Blot
Unconjugated
RTP2
Liquid
RTP2
KLH conjugated synthetic peptide between 122-151 amino acids from the central region of human RTP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit